2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Cyano-5-[(trifluoromethyl)phenyl]-2(1H)-pyridone
PC9480-1G
PC9480-500MG
APOSPC9480-1GEA
484
EUR
APOSPC9480-1G
APOSPC9480-500MG
2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile 97%
2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile
Formula:
C₁₃H₇F₃N₂O Melting Pt: 234…237 °C Storage Temperature: Ambient |
MDL Number:
MFCD00232370 CAS Number: 76053-36-6 |