1-(4-Phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
PC9481-1G PC9481-500MG
APOSPC9481-1GEA 597 EUR
APOSPC9481-1G APOSPC9481-500MG
1-(4-Phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid 97%
1-(4-Phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Formula: C₁₄H₈F₃N₃O₂S
MW: 339.3 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00665286
CAS Number: 159885-80-0