4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline
Supplier: Apollo Scientific
Synonyms: 4-Amino-3'-(trifluoromethyl)diphenyl sulphide
PC9498-1G PC9498-500MG PC9498-5G
APOSPC9498-1GEA 217 EUR
APOSPC9498-1G APOSPC9498-500MG APOSPC9498-5G
4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline
4-{[3-(Trifluoromethyl)phenyl]sulphanyl}aniline
Formula: C₁₃H₁₀F₃NS
MW: 269.29 g/mol
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00172345
CAS Number: 339104-66-4