3-Chloro-4-fluorotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-2-fluoro-5-methylbenzene, 2-Chloro-1-fluoro-4-methylbenzene
PC9527-100G PC9527-5G
APOSPC9527-100GEA 161 EUR
APOSPC9527-100G APOSPC9527-5G
3-Chloro-4-fluorotoluene 97%
3-Chloro-4-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 167…169 °C
Density: 1.18 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079730
CAS Number: 1513-25-3
UN: 1993
ADR: 3 III