2,6-Dichloro-5-fluoronicotinonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dichloro-3-cyano-5-fluoropyridine, 2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile
PC9532-25G PC9532-100G PC9532-5G
APOSPC9532-25GEA 21.9 EUR
APOSPC9532-25G APOSPC9532-100G APOSPC9532-5G
2,6-Dichloro-5-fluoronicotinonitrile 97%
2,6-Dichloro-5-fluoronicotinonitrile
Formula: C₆HCl₂FN₂
MW: 190.99 g/mol
Melting Pt: 85…93 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01863289
CAS Number: 82671-02-1
UN: 3439
ADR: 6.1 III