2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-2,3,5,6-tetrafluorobenzotrifluoride
PC9563-100G PC9563-25G PC9563-5G
APOSPC9563-100GEA 1540 EUR
APOSPC9563-100G APOSPC9563-25G APOSPC9563-5G
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol
Formula: C₇HF₇O
Boiling Pt: 91 °C/70mmHg
Melting Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00155984
CAS Number: 2787-79-3