4-(Trifluoromethyl)nicotinohydrazide 95+%
Supplier: Apollo Scientific
Warning
PC9565-1G PC9565-250MG
APOSPC9565-1GEA 847 EUR
APOSPC9565-1G APOSPC9565-250MG
4-(Trifluoromethyl)nicotinohydrazide 95+%
4-(Trifluoromethyl)nicotinohydrazide
Formula: C₇H₆F₃N₃O
Melting Pt: 157…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00101567
CAS Number: 175204-84-9