2-Chloro-3-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC9575-25G PC9575-5G PC9575-100G
APOSPC9575-25GEA 74.1 EUR
APOSPC9575-25G APOSPC9575-5G APOSPC9575-100G
2-Chloro-3-fluoroaniline 98%
2-Chloro-3-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 212…215 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03407961
CAS Number: 21397-08-0