4-Bromo-3-fluorobenzonitrile 95%
Supplier: Apollo Scientific
Warning
PC9579-100G PC9579-25G PC9579-5G
APOSPC9579-100GEA 241 EUR
APOSPC9579-100G APOSPC9579-25G APOSPC9579-5G
4-Bromo-3-fluorobenzonitrile 95%
4-Bromo-3-fluorobenzonitrile
Formula: C₇H₃BrFN
Melting Pt: 92…93 °C
Storage Temperature: Ambient
MDL Number: MFCD07782069
CAS Number: 133059-44-6
UN: 3439
ADR: 6.1,III