2-(Trifluoromethyl)phenylacetylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Ethynyl-2-(trifluoromethyl)benzene
PC9585-1G PC9585-25G PC9585-5G
APOSPC9585-1GEA 21.9 EUR
APOSPC9585-1G APOSPC9585-25G APOSPC9585-5G
2-(Trifluoromethyl)phenylacetylene 98%
2-(Trifluoromethyl)phenylacetylene
Formula: C₉H₅F₃
MW: 170.13 g/mol
Boiling Pt: 45 °C/8mmHg
Density: 1.222 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00467313
CAS Number: 704-41-6
UN: 1993
ADR: 3,III