N-Boc-4-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (4-fluorophenyl)carbamate, 4-Fluoroaniline, N-BOC protected
PC9591-1G PC9591-5G PC9591-25G
APOSPC9591-1GEA 21.9 EUR
APOSPC9591-1G APOSPC9591-5G APOSPC9591-25G
N-Boc-4-fluoroaniline 98%
N-Boc-4-fluoroaniline
Formula: C₁₁H₁₄FNO₂
MW: 211.2328 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07127707
CAS Number: 60144-53-8