1-[4-(Trifluoromethyl)-2-pyrimidinyl]-1,4-diazepane
Supplier: Apollo Scientific
Danger
Synonyms: 1-(4-(Trifluoromethyl)pyrimidin-2-yl)-1,4-diazepane, 1-[4-(Trifluoromethyl)pyrimidin-2-yl]homopiperazine
PC9612-1G PC9612-250MG PC9612-5G
APOSPC9612-1GEA 125 EUR
APOSPC9612-1G APOSPC9612-250MG APOSPC9612-5G
1-[4-(Trifluoromethyl)-2-pyrimidinyl]-1,4-diazepane
1-[4-(Trifluoromethyl)-2-pyrimidinyl]-1,4-diazepane
Formula: C₁₀H₁₃F₃N₄
MW: 246.24 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00728997
CAS Number: 238403-48-0
UN: 3263
ADR: 8,III