2-Amino-6-(trifluoromethyl)pyridine 97+%
Supplier: Apollo Scientific
Danger
PC9615-100G PC9615-25G PC9615-5G
APOSPC9615-100GEA 101 EUR
APOSPC9615-100G APOSPC9615-25G APOSPC9615-5G
2-Amino-6-(trifluoromethyl)pyridine 97+%
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,III