3,4,5-Trifluorobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: 3,4,5-Trifluorobenzene-1-sulphonyl chloride
PC9616-100G PC9616-25G PC9616-5G
APOSPC9616-100GEA 313 EUR
APOSPC9616-100G APOSPC9616-25G APOSPC9616-5G
3,4,5-Trifluorobenzenesulphonyl chloride 95%
3,4,5-Trifluorobenzenesulphonyl chloride
Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Boiling Pt: 76…77 °C
Density: 1.658 g/cm³ (25 °C)
Flash Pt: 23.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03094559
CAS Number: 351003-43-5
UN: 2924
ADR: 3+8 III