4-Chloro-2-fluorobenzenesulphonyl chloride 96%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Chloro-2-fluorobenzene-1-sulphonyl chloride
PC9617-5G PC9617-1G
APOSPC9617-5GEA 80 EUR
APOSPC9617-5G APOSPC9617-1G
4-Chloro-2-fluorobenzenesulphonyl chloride 96%
4-Chloro-2-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Melting Pt: 34…38 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094560
CAS Number: 141337-26-0
UN: 3261
ADR: 8,II