1-Bromo-4-iodo-1,1,2,2-tetrafluorobutane
Supplier: Apollo Scientific
Warning
PC9624-1G PC9624-5G
APOSPC9624-1GEA 139 EUR
APOSPC9624-1G APOSPC9624-5G
1-Bromo-4-iodo-1,1,2,2-tetrafluorobutane
1-Bromo-4-iodo-1,1,2,2-tetrafluorobutane
Formula: C₄H₄BrF₄I
Flash Pt: 110 °C
Storage Temperature: Ambient
CAS Number: 129587-49-1