2,2-Difluoroethylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2-Difluoroethanamine hydrochloride
PC9627-1G PC9627-5G
APOSPC9627-1GEA 23.2 EUR
APOSPC9627-1G APOSPC9627-5G
2,2-Difluoroethylamine hydrochloride 98%
2,2-Difluoroethylamine hydrochloride
Formula: C₂H₆ClF₂N
MW: 117.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038283
CAS Number: 79667-91-7