2,3-Dibromohexafluoro-2-butene 97%
Supplier: Apollo Scientific
PC9654-25G PC9654-5G
APOSPC9654-25GEA 579 EUR
APOSPC9654-25G APOSPC9654-5G
2,3-Dibromohexafluoro-2-butene 97%
2,3-Dibromohexafluoro-2-butene
Formula: C₄Br₂F₆
MW: 321.85 g/mol
Boiling Pt: 104…106 °C
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01862090
CAS Number: 384-51-0