4-Fluoroindole
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-1H-indole
PC9665-1G PC9665-5G PC9665-10G
APOSPC9665-1GEA 21.9 EUR
APOSPC9665-1G APOSPC9665-5G APOSPC9665-10G
4-Fluoroindole
4-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 90 °C/0.4mmHg
Melting Pt: 29…33 °C
Storage Temperature: Freezer
MDL Number: MFCD00055992
CAS Number: 387-43-9