2,3,5,6-Tetrafluoro-4-hydroxypyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,5,6-Tetrafluoropyridin-4-ol, 4-Hydroxy-2,3,5,6-tetrafluoropyridine
PC9677-1G PC9677-250MG
APOSPC9677-1GEA 109 EUR
APOSPC9677-1G APOSPC9677-250MG
2,3,5,6-Tetrafluoro-4-hydroxypyridine
2,3,5,6-Tetrafluoro-4-hydroxypyridine
Formula: C₅HF₄NO
MW: 167.063 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00981885
CAS Number: 2693-66-5