4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Mercapto-6-(trifluoromethyl)-4-pyrimidinol, 4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 4-Hydroxy-2-mercapto-6-(trifluoromethyl)pyrimidine
PC9778-1G PC9778-25G
APOSPC9778-1GEA 21.9 EUR
APOSPC9778-1G APOSPC9778-25G
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol
Formula: C₅H₃F₃N₂OS
MW: 196.10 g/mol
Melting Pt: 246…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00112978
CAS Number: 368-54-7