Methyl undecafluorohexanoate 97%
Supplier: Apollo Scientific
Danger
Synonyms: Undecafluorohexanoic Acid Methyl Ester, Perfluorohexanoic Acid Methyl Ester, Methyl 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoate, Methyl Perfluorohexanoate
PC9788-25G PC9788-5G
APOSPC9788-25GEA 149 EUR
APOSPC9788-25G APOSPC9788-5G
Methyl undecafluorohexanoate 97%
Methyl undecafluorohexanoate
Formula: C₇H₃F₁₁O₂
MW: 328.08 g/mol
Boiling Pt: 121 °C
Density: 1.579 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06200802
CAS Number: 424-18-0
UN: 1993
ADR: 3 II