2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
Supplier: Apollo Scientific
PC9790-25G PC9790-5G PC9790-1G
APOSPC9790-25GEA 1260 EUR
APOSPC9790-25G APOSPC9790-5G APOSPC9790-1G
2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
2-Quinolin-2-yl-1-[4-(trifluoromethyl)phenyl]ethanone
Formula: C₁₈H₁₂F₃NO
MW: 315.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037910
CAS Number: 283597-72-8