2-Hydrazino-3-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydrazinyl-3-(trifluoromethyl)pyridine
PC9803-1G PC9803-5G
APOSPC9803-1GEA 34.8 EUR
APOSPC9803-1G APOSPC9803-5G
2-Hydrazino-3-(trifluoromethyl)pyridine
2-Hydrazino-3-(trifluoromethyl)pyridine
Formula: C₆H₆F₃N₃
MW: 177.13 g/mol
Melting Pt: 64…65 °C
Density: 0.936 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01320804
CAS Number: 89570-83-2