2-Bromo-5'-fluoro-2'-hydroxyacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2-Bromo-1-(5-fluoro-2-hydroxyphenyl)ethanone, 5-Fluoro-2-hydroxyphenacyl bromide
PC9804-1G PC9804-5G PC9804-25G
APOSPC9804-1GEA 29 EUR
APOSPC9804-1G APOSPC9804-5G APOSPC9804-25G
2-Bromo-5'-fluoro-2'-hydroxyacetophenone
2-Bromo-5'-fluoro-2'-hydroxyacetophenone
Formula: C₈H₆BrFO₂
MW: 233.0345 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD03094564
CAS Number: 126581-65-5
UN: 3335
ADR: 9,III