Methyl-2-amino-5-fluorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-Amino-5-fluorobenzoate, 5-Fluoroanthranilic acid methyll ester, Methyl 5-Fluoroanthranilate, 2-Amino-5-fluorobenzoic acid methyll ester, Methyl 2-amino-5-fluorobenzoate
PC9817-100G
APOSPC9817-100GEA 194 EUR
APOSPC9817-100G
Methyl-2-amino-5-fluorobenzoate 98%
Methyl 2-amino-5-fluorobenzoate
Formula: C₈H₈FNO₂
MW: 169.16 g/mol
Boiling Pt: 100…102 °C/0.5mmHg
Melting Pt: 34…40 °C
Density: 1.26 g/cm³ (25 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD03791308
CAS Number: 319-24-4