2-Quinolin-4-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
Supplier: Apollo Scientific
Warning
PC9820-1G PC9820-5G PC9820-25G
APOSPC9820-1GEA 163 EUR
APOSPC9820-1G APOSPC9820-5G APOSPC9820-25G
2-Quinolin-4-yl-1-[4-(trifluoromethyl)phenyl]ethanone 95+%
2-Quinolin-4-yl-1-[4-(trifluoromethyl)phenyl]ethanone
Formula: C₁₈H₁₂F₃NO
MW: 315.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04037911
CAS Number: 476472-22-7