3,3-Difluoropiperidine hydrochloride
Supplier: Apollo Scientific
Warning
PC9831-1G PC9831-5G
APOSPC9831-1GEA 89.9 EUR
APOSPC9831-1G APOSPC9831-5G
3,3-Difluoropiperidine hydrochloride
3,3-Difluoropiperidine hydrochloride
Formula: C5H10ClF2N
MW: 157.59 g/mol
Melting Pt: 236…239 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD03452800
CAS Number: 496807-97-7