4-Nitro-1,2-bis(trifluoromethyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Bis(trifluoromethyl)nitrobenzene
PC9833-1G PC9833-5G
APOSPC9833-1GEA 145 EUR
APOSPC9833-1G APOSPC9833-5G
4-Nitro-1,2-bis(trifluoromethyl)benzene
4-Nitro-1,2-bis(trifluoromethyl)benzene
Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
Boiling Pt: 103…104 °C/18mmHg
Melting Pt: 23…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00728841
CAS Number: 1978-20-7