1-(Trifluoromethyl)cyclobutanecarboxylic acid
Supplier: Apollo Scientific
Danger
PC9836-1G PC9836-5G
APOSPC9836-1GEA 87.3 EUR
APOSPC9836-1G APOSPC9836-5G
1-(Trifluoromethyl)cyclobutanecarboxylic acid
1-(Trifluoromethyl)cyclobutanecarboxylic acid
Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 76 °C/14mmHg
Melting Pt: 24…26 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425197
CAS Number: 277756-45-3
UN: 2811
ADR: 6.1,III