Methyl-2,3-difluorobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2,3-difluorobenzoate
PC9846-10G PC9846-25G PC9846-5G
APOSPC9846-10GEA 27.7 EUR
APOSPC9846-10G APOSPC9846-25G APOSPC9846-5G
Methyl-2,3-difluorobenzoate
Methyl 2,3-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Boiling Pt: 94 °C/ 15mmHg
Storage Temperature: Ambient
MDL Number: MFCD06203674
CAS Number: 18355-74-3