2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane 97%
Supplier: Apollo Scientific
Synonyms: 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-Icosafluoro-5,8,11-tris(trifluoromethyl)-3,6,9,12-tetraoxapentadecane
PC9855-25G PC9855-5G PC9855-1G
APOSPC9855-25GEA 183 EUR
APOSPC9855-25G APOSPC9855-5G APOSPC9855-1G
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane 97%
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane
Formula: C₁₄HF₂₉O₄
MW: 784.12 g/mol
Boiling Pt: 192…195 °C
Melting Pt: –115 °C
Density: 1.723 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054717
CAS Number: 26738-51-2