Tetrafluorobenzene-1,3-diol 97%
Supplier: Apollo Scientific
Danger
PC9866-5G PC9866-500MG
APOSPC9866-5GEA 277 EUR
APOSPC9866-5G APOSPC9866-500MG
Tetrafluorobenzene-1,3-diol 97%
Tetrafluorobenzene-1,3-diol
Formula: C₆H₂F₄O₂
MW: 182.08 g/mol
Boiling Pt: 217…218 °C
Melting Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00040202
CAS Number: 16840-25-8