4-(Trifluoromethyl)-1H-imidazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Trifluoromethyl)imidazole
PC9869-5G PC9869-1G PC9869-25G
APOSPC9869-5GEA 116 EUR
APOSPC9869-5G APOSPC9869-1G APOSPC9869-25G
4-(Trifluoromethyl)-1H-imidazole 97%
4-(Trifluoromethyl)-1H-imidazole
Formula: C₄H₃F₃N₂
MW: 136.08 g/mol
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD08458860
CAS Number: 33468-69-8