Bis(1H,1H,2H,2H-perfluorooctyl)maleate
Supplier: Apollo Scientific
Warning
PC9892-25G PC9892-5G
APOSPC9892-25GEA 0 EUR
APOSPC9892-25G APOSPC9892-5G
Bis(1H,1H,2H,2H-perfluorooctyl)maleate
Bis(1H,1H,2H,2H-perfluorooctyl)maleate
Formula: C₂₀H₁₀F₂₆O₄
MW: 808.26 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD04038264
CAS Number: 55003-96-8