2-Bromo-6-fluoroaniline 99%
Supplier: Apollo Scientific
Warning
PC9902-100G PC9902-25G PC9902-5G PC9902-1KG
APOSPC9902-100GEA 132 EUR
APOSPC9902-100G APOSPC9902-25G APOSPC9902-5G APOSPC9902-1KG
2-Bromo-6-fluoroaniline 99%
2-Bromo-6-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Density: 1.674 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01310982
CAS Number: 65896-11-9