3-Bromo-4-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC9908-5G PC9908-100G PC9908-25G
APOSPC9908-5GEA 14.7 EUR
APOSPC9908-5G APOSPC9908-100G APOSPC9908-25G
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 124 °C/11mmHg
Melting Pt: 29…36 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD03788559
CAS Number: 656-64-4
UN: 2811
ADR: 6.1,III