3-Hydroxy-5-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
Warning
PC9912-10G PC9912-1G PC9912-5G
APOSPC9912-10GEA 232 EUR
APOSPC9912-10G APOSPC9912-1G APOSPC9912-5G
3-Hydroxy-5-(trifluoromethyl)benzoic acid
3-Hydroxy-5-(trifluoromethyl)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 191…193 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368783
CAS Number: 328-69-8