4-(Bromomethyl)-3-fluorobenzonitrile 97%
Supplier: Apollo Scientific
Danger
PC9915-1G PC9915-25G PC9915-5G
APOSPC9915-1GEA 21.9 EUR
APOSPC9915-1G APOSPC9915-25G APOSPC9915-5G
4-(Bromomethyl)-3-fluorobenzonitrile 97%
4-(Bromomethyl)-3-fluorobenzonitrile
Formula: C₈H₅BrFN
MW: 214.04 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07368341
CAS Number: 105942-09-4
UN: 3439
ADR: 6.1 III