1-Bromo-3-fluoro-2-nitrobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-6-fluoronitrobenzene
PC9918-25G PC9918-5G PC9918-100G
APOSPC9918-25GEA 92.8 EUR
APOSPC9918-25G APOSPC9918-5G APOSPC9918-100G
1-Bromo-3-fluoro-2-nitrobenzene 99%
1-Bromo-3-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 36…37 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD07368788
CAS Number: 886762-70-5