α-Bromo-2-(difluoromethoxy)toluene 98%
Supplier: Apollo Scientific
Danger
Synonyms: o-(Difluoromethoxy)benzyl bromide, 1-(Bromomethyl)-2-(difluoromethoxy)benzene, 2-(Difluoromethoxy)benzyl bromide
PC9921-100G PC9921-25G PC9921-5G
APOSPC9921-100GEA 397 EUR
APOSPC9921-100G APOSPC9921-25G APOSPC9921-5G
α-Bromo-2-(difluoromethoxy)toluene 98%
α-Bromo-2-(difluoromethoxy)toluene
Formula: C₈H₇BrF₂O
MW: 237.04 g/mol
Boiling Pt: 222 °C
Melting Pt: 13
Density: 1.555 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: 8 II