4'-Bromo-2,2,2-trifluoroacetophenone 99%
Supplier: Apollo Scientific
Warning
PC9923-100G PC9923-25G PC9923-5G
APOSPC9923-100GEA 253 EUR
APOSPC9923-100G APOSPC9923-25G APOSPC9923-5G
4'-Bromo-2,2,2-trifluoroacetophenone 99%
4'-Bromo-2,2,2-trifluoroacetophenone
Formula: C₈H₄BrF₃O
Boiling Pt: 95 °C/4mmHg
Melting Pt: 26…30 °C
Density: 1.662 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00191862
CAS Number: 16184-89-7