5-Bromo-2,4-difluoroaniline 98%
Supplier: Apollo Scientific
Danger
PC9925-25G PC9925-5G
APOSPC9925-25GEA 43.5 EUR
APOSPC9925-25G APOSPC9925-5G
5-Bromo-2,4-difluoroaniline 98%
5-Bromo-2,4-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.0000 g/mol
Melting Pt: 25…29 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426368
CAS Number: 452-92-6
UN: 2811
ADR: 6.1 III