3-Fluoro-4-methylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Fluoro-4-picoline
PC9930-1G PC9930-5G PC9930-10G
APOSPC9930-1GEA 21.9 EUR
APOSPC9930-1G APOSPC9930-5G APOSPC9930-10G
3-Fluoro-4-methylpyridine
3-Fluoro-4-methylpyridine
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 135.6 °C/748mmHg
Density: 1.08 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04114226
CAS Number: 399-88-2