5-Fluorocytidine
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one
PC9941-1G PC9941-250MG
APOSPC9941-1GEA 24.8 EUR
APOSPC9941-1G APOSPC9941-250MG
5-Fluorocytidine
5-Fluorocytidine
Formula: C₉H₁₂FN₃O₅
Melting Pt: 196…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00210953
CAS Number: 2341-22-2