3,5-Difluorobenzenesulphonamide
Supplier: Apollo Scientific
Warning
PC9957-1G PC9957-5G PC9957-25G
APOSPC9957-1GEA 21.9 EUR
APOSPC9957-1G APOSPC9957-5G APOSPC9957-25G
3,5-Difluorobenzenesulphonamide
3,5-Difluorobenzenesulphonamide
Formula: C₆H₅F₂NO₂S
MW: 193.17 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD02091379
CAS Number: 140480-89-3