3,5-Difluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3,5-Difluorobenzene-1-sulphonyl chloride
PC9972-5G PC9972-25G PC9972-100G
APOSPC9972-5GEA 0 EUR
APOSPC9972-5G APOSPC9972-25G APOSPC9972-100G
3,5-Difluorobenzenesulphonyl chloride 98%
3,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
Melting Pt: 57…59 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,II