5-Chloro-2,4,6-trifluoropyrimidine 95%
Supplier: Apollo Scientific
Warning
PC9980-1G PC9980-5G
APOSPC9980-1GEA 21.9 EUR
APOSPC9980-1G APOSPC9980-5G
5-Chloro-2,4,6-trifluoropyrimidine 95%
5-Chloro-2,4,6-trifluoropyrimidine
Formula: C₄ClF₃N₂
MW: 168.50 g/mol
Boiling Pt: 116 °C
Density: 1.626 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00191933
CAS Number: 697-83-6
EINECS: 211-807-4