4-Chloro-3-fluorotoluene
Supplier: Apollo Scientific
Warning
PC9987-25G PC9987-5G PC9987-100G
APOSPC9987-25GEA 29 EUR
APOSPC9987-25G APOSPC9987-5G APOSPC9987-100G
4-Chloro-3-fluorotoluene
4-Chloro-3-fluorotoluene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 157…158 °C
Density: 1.2 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00143295
CAS Number: 5527-94-6
UN: 1993
ADR: 3 III