Photometric tube tests, NANOCOLOR® cationic Surfactants
Supplier: Macherey-Nagel
NANOCOLOR®
MANA985034EA 107 EUR
MANA985034
Photometric tube tests, NANOCOLOR® cationic Surfactants
Environmental Test Kits
A pre-dosed NANOCOLOR tube test for photometric determination of cationic surfactants in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes